Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204221.1 | NP_001191150.1 | 2 | 268 | ||
NM_020394.4 | NP_065127.4 | 2 | 268 | |||
NR_037892.1 | 2 | 268 | ||||
GenBank mRNA | BC023527.2 | - | 2 | 176 | ||
BC041082.1 | - | 2 | 205 | |||
BC055096.1 | - | 2 | 232 | |||
CN394881.1 | - | 2 | 200 | |||
DA809929.1 | - | 2 | 268 |
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 2 | 300 | |||
GenBank mRNA | AK126366.1 | - | 2 | 298 | ||
AL524527.3 | - | 2 | 223 |