Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 6 | 2461 | ||
NM_020746.4 | NP_065797.2 | 7 | 2636 | |||
NR_037921.1 | 6 | 2463 | ||||
GenBank mRNA | AB033097.1 | - | 6 | 2542 | ||
AB097003.1 | - | 8 | 2000 | |||
AB232371.1 | - | 6 | 2536 | |||
AK023799.1 | - | 7 | 2591 | |||
AK123956.1 | - | 4 | 458 | |||
AL833736.1 | - | 1 | 2691 | |||
BC044952.1 | - | 7 | 2570 | 5751684 | ||
BX649084.1 | - | 6 | 2422 |