Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 5 | 1979 | ||
XM_005263163.4 | XP_005263220.1 | 4 | 6433 | |||
XM_005263164.4 | XP_005263221.1 | 4 | 991 | |||
XM_005263165.4 | XP_005263222.1 | 4 | 2417 | |||
XM_006714282.3 | XP_006714345.1 | 5 | 1134 | |||
XM_011532157.2 | XP_011530459.2 | 4 | 1821 | |||
XM_011532158.2 | XP_011530460.1 | 6 | 1780 | |||
XM_011532159.1 | XP_011530461.1 | 5 | 1969 | |||
XM_011532160.1 | XP_011530462.1 | 3 | 981 | |||
XM_011532161.2 | XP_011530463.1 | 4 | 1159 | |||
XM_017008488.1 | XP_016863977.1 | 5 | 1137 | |||
XM_017008489.1 | XP_016863978.1 | 5 | 1038 | |||
GenBank mRNA | AB055660.1 | - | 5 | 1980 | ||
AK128677.1 | - | 1 | 481 | |||
AK304103.1 | - | 4 | 775 |