Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 8 | 6002 | ||
XM_005263163.4 | XP_005263220.1 | 7 | 10456 | |||
XM_005263164.4 | XP_005263221.1 | 7 | 5014 | |||
XM_005263165.4 | XP_005263222.1 | 7 | 6440 | |||
XM_006714282.3 | XP_006714345.1 | 8 | 5157 | |||
XM_011532157.2 | XP_011530459.2 | 7 | 5844 | |||
XM_011532158.2 | XP_011530460.1 | 9 | 5803 | |||
XM_011532159.1 | XP_011530461.1 | 8 | 5992 | |||
XM_011532160.1 | XP_011530462.1 | 6 | 5004 | |||
XM_011532161.2 | XP_011530463.1 | 7 | 5182 | |||
XM_017008488.1 | XP_016863977.1 | 8 | 5160 | |||
XM_017008489.1 | XP_016863978.1 | 8 | 5061 | |||
GenBank mRNA | AB040914.1 | - | 4 | 3201 | ||
AB055660.1 | - | 8 | 6003 | |||
AK027744.1 | - | 5 | 939 | |||
AK128677.1 | - | 4 | 4504 | |||
BC007291.1 | - | 4 | 1363 |