Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195194.1 | NP_001182123.1 | 3 | 304 | ||
NM_001251964.1 | NP_001238893.1 | 2 | 2835 | |||
NM_022112.2 | NP_071395.2 | 4 | 982 | |||
XM_017018113.1 | XP_016873602.1 | 4 | 1747 | |||
XM_017018114.1 | XP_016873603.1 | 4 | 1741 | |||
XM_017018116.1 | XP_016873605.1 | 4 | 5012 | |||
GenBank mRNA | AB045830.1 | - | 4 | 510 | ||
AB045832.1 | - | 2 | 2363 |