Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022371.3 | NP_071766.2 | 5 | 1045 | ||
GenBank mRNA | AJ299403.1 | - | 5 | 934 | ||
AJ299441.1 | - | 5 | 934 | |||
AK025998.1 | - | 5 | 924 | |||
AK130132.1 | - | 3 | 364 | |||
AK299966.1 | - | 4 | 642 | |||
AK316530.1 | - | 4 | 654 | |||
BC001085.1 | - | 5 | 943 | 2959819 | ||
BC007571.1 | - | 5 | 934 | 3029304 | ||
BC011746.1 | - | 5 | 930 | 3343691 | ||
BC018292.1 | - | 5 | 935 | 3457259 | ||
BC088368.1 | - | 4 | 672 | |||
CR457354.1 | - | 5 | 893 |