Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022491.2 | NP_071936.2 | 11 | 966 | ||
XM_011538671.2 | XP_011536973.1 | 12 | 2043 | |||
XM_017019817.1 | XP_016875306.1 | 10 | 850 | |||
XM_017019818.1 | XP_016875307.1 | 13 | 1198 | |||
XM_017019819.1 | XP_016875308.1 | 12 | 1060 | |||
XM_017019820.1 | XP_016875309.1 | 13 | 1092 | |||
GenBank mRNA | AK023801.1 | - | 6 | 442 | ||
AK024460.1 | - | 11 | 949 | |||
BC030252.1 | - | 5 | 339 | |||
BC093990.1 | - | 6 | 396 | 7939835 | ||
BC098404.1 | - | 11 | 989 | 30332266 | ||
BC112000.1 | - | 6 | 396 | |||
BX647687.1 | - | 12 | 924 |