Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022773.2 | NP_073610.2 | 11 | 1625 | ||
NR_033645.1 | 12 | 1754 | ||||
NR_033646.1 | 13 | 1932 | ||||
XM_006720928.2 | XP_006720991.1 | 11 | 1898 | |||
XM_006720929.3 | XP_006720992.1 | 12 | 1779 | |||
XM_011522617.2 | XP_011520919.1 | 9 | 1274 | |||
XM_017023576.1 | XP_016879065.1 | 10 | 1389 | |||
XM_017023577.1 | XP_016879066.1 | 12 | 2054 | |||
XM_017023578.1 | XP_016879067.1 | 10 | 1474 | |||
XM_017023579.1 | XP_016879068.1 | 10 | 1482 | |||
XM_017023580.1 | XP_016879069.1 | 13 | 2204 | |||
GenBank mRNA | AB075873.1 | - | 11 | 1609 | ||
AK022743.1 | - | 11 | 1623 | |||
AK025955.1 | - | 11 | 1557 | |||
AK093050.1 | - | 8 | 1132 | |||
AK125201.1 | - | 12 | 1756 | |||
AK225065.1 | - | 11 | 1623 | |||
AK225406.1 | - | 11 | 1610 | |||
AL832177.1 | - | 5 | 3243 | |||
BC010738.1 | - | 10 | 1390 | |||
BC081535.1 | - | 1 | 1334 |