Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261392.1 | NP_001248321.1 | 14 | 1248 | ||
NM_001261394.1 | NP_001248323.1 | 14 | 1176 | |||
NM_024894.3 | NP_079170.2 | 15 | 1326 | |||
NR_048552.1 | 14 | 1280 | ||||
XM_011510398.2 | XP_011508700.1 | 14 | 1193 | |||
XM_011510399.2 | XP_011508701.1 | 13 | 1131 | |||
XM_017004990.1 | XP_016860479.1 | 11 | 851 | |||
XM_017004991.1 | XP_016860480.1 | 4 | 303 | |||
GenBank mRNA | AK024137.1 | - | 14 | 1161 | ||
AK290680.1 | - | 15 | 1290 | |||
AK297169.1 | - | 14 | 1236 | |||
AK299617.1 | - | 14 | 1268 | |||
BC005125.2 | - | 15 | 1306 | 3841643 | ||
BM021448.1 | - | 2 | 90 |