Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261392.1 | NP_001248321.1 | 5 | 482 | ||
NM_001261394.1 | NP_001248323.1 | 6 | 560 | |||
NM_024894.3 | NP_079170.2 | 6 | 560 | |||
NR_048552.1 | 5 | 514 | ||||
XM_011510400.2 | XP_011508702.1 | 6 | 548 | |||
XM_011510402.2 | XP_011508704.1 | 6 | 548 | |||
XM_017004990.1 | XP_016860479.1 | 4 | 302 | |||
GenBank mRNA | AK024137.1 | - | 6 | 545 | ||
AK290680.1 | - | 6 | 524 | |||
AK297169.1 | - | 5 | 470 | |||
AK299617.1 | - | 5 | 502 | |||
BC005125.2 | - | 6 | 540 | 3841643 | ||
BC037841.1 | - | 1 | 1620 | |||
BG390065.1 | - | 1 | 15 | |||
DC346574.1 | - | 6 | 560 |