Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 1 | 79 | ||
NM_001171944.1 | NP_001165415.1 | 2 | 138 | |||
NM_001171945.1 | NP_001165416.1 | 3 | 358 | |||
NM_001171946.1 | NP_001165417.1 | 2 | 126 | |||
GenBank mRNA | AB018353.1 | - | 2 | 96 | ||
AB648918.1 | - | 1 | 59 | |||
AK022469.1 | - | 2 | 106 | |||
AK302896.1 | - | 2 | 110 | |||
AK309120.1 | - | 3 | 330 | |||
BC013613.1 | - | 2 | 126 | |||
BC142707.1 | - | 1 | 89 | 40148817 | ||
BC152418.1 | - | 1 | 96 | 8862528 | ||
BX538211.1 | - | 2 | 107 | |||
DA112466.1 | - | 2 | 138 |