Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308189.1 | NP_001295118.1 | 9 | 3532 | ||
NM_001308191.1 | NP_001295120.1 | 8 | 3508 | |||
NM_001308192.1 | NP_001295121.1 | 9 | 1161 | |||
NM_001308193.1 | NP_001295122.1 | 8 | 1110 | |||
NM_030627.3 | NP_085130.2 | 10 | 3583 | |||
XM_005265994.1 | XP_005266051.1 | 9 | 3559 | |||
GenBank mRNA | AB051460.1 | - | 9 | 2194 | ||
AB586689.1 | - | 5 | 3567 | |||
BC036899.1 | - | 8 | 1094 | |||
BC117150.1 | - | 10 | 2571 | 40125701 | ||
BC143958.1 | - | 9 | 2520 | 9052476 | ||
BC143959.1 | - | 8 | 2496 | 9052477 | ||
BX538213.1 | - | 10 | 2725 |