Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032134.2 | NP_115510.1 | 18, 19 | 5164 | ||
NR_130649.1 | 18, 19 | 2255 | ||||
GenBank mRNA | AK058102.1 | - | 10, 11 | 1306 | ||
AL136774.1 | - | 18, 19 | 5164 | |||
BC131559.1 | - | 18, 19 | 2140 |