Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016287.3 | NP_057371.2 | 4 | 364 | ||
XM_005245875.4 | XP_005245932.1 | 4 | 939 | |||
XM_005245876.3 | XP_005245933.1 | 4 | 472 | |||
XM_005245877.4 | XP_005245934.1 | 4 | 800 | |||
XM_005245878.4 | XP_005245935.1 | 4 | 736 | |||
XM_005245879.4 | XP_005245936.1 | 3 | 263 | |||
XM_011541532.1 | XP_011539834.1 | 4 | 456 | |||
XM_011541533.1 | XP_011539835.1 | 4 | 653 | |||
XM_011541534.2 | XP_011539836.1 | 3 | 273 | |||
XM_017001393.1 | XP_016856882.1 | 4 | 419 | |||
XM_017001394.1 | XP_016856883.1 | 4 | 784 | |||
GenBank mRNA | AF113534.1 | - | 3 | 294 | ||
AK023129.1 | - | 4 | 364 | |||
AK291252.1 | - | 4 | 342 | |||
AK304065.1 | - | 3 | 240 | |||
BC032139.2 | - | 4 | 451 | |||
BC046170.1 | - | 4 | 399 | |||
CK000759.1 | - | 1 | 26 |