Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033127.2 | NP_149118.2 | 15 | 1913 | ||
GenBank mRNA | AB063357.1 | - | 15 | 1913 | ||
AK090971.1 | - | 1 | 15 | |||
AK098627.1 | - | 4 | 533 | |||
AK314585.1 | - | 14 | 1917 | |||
EF125213.1 | - | 14 | 1823 |