Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134222.1 | NP_001127694.1 | 3 | 1399 | ||
NM_033267.4 | NP_150366.1 | 3 | 1399 | |||
XM_011513979.2 | XP_011512281.1 | 3 | 1326 | |||
GenBank mRNA | AB188492.1 | - | 3 | 1402 | ||
AY335940.1 | - | 3 | 1167 | |||
BC065189.1 | - | 3 | 1406 | 6422941 |