Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 3, 4 | 585 | ||
XM_005251318.3 | XP_005251375.1 | 2, 3 | 374 | |||
XM_005251319.4 | XP_005251376.1 | 3, 4 | 454 | |||
XM_005251320.3 | XP_005251377.1 | 2, 3 | 246 | |||
XM_005251322.3 | XP_005251379.1 | 2, 3 | 254 | |||
XM_017013920.1 | XP_016869409.1 | 3, 4 | 491 | |||
XM_017013921.1 | XP_016869410.1 | 3, 4 | 491 | |||
XM_017013922.1 | XP_016869411.1 | 3, 4 | 491 | |||
XM_017013923.1 | XP_016869412.1 | 2, 3 | 318 | |||
XM_017013924.1 | XP_016869413.1 | 3, 4 | 491 | |||
XM_017013925.1 | XP_016869414.1 | 2, 3 | 273 | |||
XM_017013926.1 | XP_016869415.1 | 2, 3 | 256 | |||
XM_017013927.1 | XP_016869416.1 | 2, 3 | 242 | |||
XM_017013928.1 | XP_016869417.1 | 2, 3 | 122 | |||
GenBank mRNA | AB051551.1 | - | 2, 3 | 1206 | ||
AK056185.1 | - | 2, 3 | 548 | |||
AK300234.1 | - | 2, 3 | 380 | |||
AK300874.1 | - | 2, 3 | 229 | |||
AK302648.1 | - | 3, 4 | 435 | |||
AK307350.1 | - | 2, 3 | 229 | |||
AK307443.1 | - | 2, 3 | 227 | |||
AK307636.1 | - | 2, 3 | 219 | |||
BC030701.2 | - | 3, 4 | 458 | 4837403 |