Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030771.1 | NP_110398.1 | 3 | 640 | ||
NM_080654.2 | NP_542385.1 | 3 | 640 | |||
GenBank mRNA | AF155108.1 | - | 3 | 1266 | ||
AF382034.1 | - | 3 | 528 | |||
AK308494.1 | - | 4 | 588 | |||
AK313358.1 | - | 3 | 607 | |||
AL555720.3 | - | 3 | 624 | |||
AY598334.1 | - | 3 | 586 | |||
BC008496.1 | - | 3 | 608 | 4276656 |