Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130777.2 | NP_570133.1 | 4 | 470 | ||
XM_011530836.2 | XP_011529138.1 | 4 | 498 | |||
GenBank mRNA | AJ318880.1 | - | 4 | 445 | ||
BC009232.2 | - | 4 | 450 | 3960352 | ||
BC066311.1 | - | 4 | 452 | 30330532 |