Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DQX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133637.2 | NP_598376.2 | 5 | 1128 | ||
XM_011532644.1 | XP_011530946.1 | 4 | 807 | |||
XM_011532645.1 | XP_011530947.1 | 2 | 416 | |||
XM_011532646.2 | XP_011530948.1 | 5 | 1128 | |||
XM_017003520.1 | XP_016859009.1 | 3 | 581 | |||
GenBank mRNA | AK074337.1 | - | 5 | 1035 | ||
AK092420.1 | - | 5 | 1129 | |||
BC075018.2 | - | 3 | 655 | 30915408 |