Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144688.4 | NP_653289.3 | 15, 16 | 1482 | ||
XM_005258549.3 | XP_005258606.1 | 15, 16 | 1482 | |||
XM_011526488.2 | XP_011524790.1 | 15, 16 | 1482 | |||
XM_011526489.2 | XP_011524791.1 | 15, 16 | 3382 | |||
XM_011526490.2 | XP_011524792.1 | 15, 16 | 1476 | |||
XM_011526494.2 | XP_011524796.1 | 16, 17 | 1455 | |||
XM_011526495.2 | XP_011524797.1 | 16, 17 | 1491 | |||
XM_011526496.2 | XP_011524798.1 | 14, 15 | 1343 | |||
XM_011526497.2 | XP_011524799.1 | 15, 16 | 1441 | |||
XM_017026333.1 | XP_016881822.1 | 14, 15 | 1615 | |||
GenBank mRNA | AK057220.1 | - | 15, 16 | 1451 | ||
BC029811.1 | - | 15, 16 | 1401 | 5171712 |