Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOSPHO2-KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199290.1 | NP_001186219.1 | 4 | 1316 | ||
GenBank mRNA | AK090653.1 | - | 4 | 1279 | ||
AX746559.1 | - | 4 | 1279 |
Gene Symbol: KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144711.5 | NP_653312.2 | 2 | 1185 | ||
GenBank mRNA | AK095131.1 | - | 761 | |||
BC010437.1 | - | 1019 | 3354344 |