Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317037.1 | NP_001303966.1 | 8 | 1140 | ||
NM_144965.2 | NP_659402.1 | 8 | 1179 | |||
XM_005251733.1 | XP_005251790.1 | 7 | 991 | |||
XM_005251734.1 | XP_005251791.1 | 8 | 1164 | |||
XM_006716970.1 | XP_006717033.1 | 8 | 1139 | |||
XM_006716971.1 | XP_006717034.1 | 7 | 966 | |||
XM_011518283.1 | XP_011516585.1 | 8 | 1177 | |||
XM_011518284.1 | XP_011516586.1 | 7 | 1035 | |||
XM_011518285.1 | XP_011516587.1 | 8 | 1177 | |||
XM_011518288.1 | XP_011516590.1 | 7 | 1035 | |||
XM_011518289.2 | XP_011516591.1 | 8 | 1063 | |||
XM_011518290.1 | XP_011516592.1 | 7 | 1010 | |||
XM_011518291.1 | XP_011516593.1 | 8 | 1164 | |||
XM_011518292.1 | XP_011516594.1 | 7 | 1035 | |||
XM_011518293.1 | XP_011516595.1 | 8 | 1177 | |||
XM_011518294.2 | XP_011516596.1 | 8 | 1177 | |||
XM_011518295.1 | XP_011516597.1 | 8 | 1177 | |||
XM_011518297.1 | XP_011516599.1 | 8 | 1177 | |||
XM_011518298.2 | XP_011516600.1 | 8 | 1177 | |||
GenBank mRNA | AK057342.1 | - | 8 | 1164 | ||
AK074105.1 | - | 3 | 557 | |||
AK128011.1 | - | 5 | 1107 | |||
AK294862.1 | - | 7 | 1035 | |||
AK302422.1 | - | 7 | 991 | |||
AK302737.1 | - | 8 | 1125 | |||
BC031281.1 | - | 8 | 1177 | 5295890 |