Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317037.1 | NP_001303966.1 | 14 | 2047 | ||
NM_144965.2 | NP_659402.1 | 14 | 2086 | |||
XM_005251733.1 | XP_005251790.1 | 13 | 1898 | |||
XM_005251734.1 | XP_005251791.1 | 13 | 1929 | |||
XM_006716970.1 | XP_006717033.1 | 14 | 2046 | |||
XM_006716971.1 | XP_006717034.1 | 13 | 1873 | |||
XM_011518283.1 | XP_011516585.1 | 13 | 1949 | |||
XM_011518284.1 | XP_011516586.1 | 13 | 1942 | |||
XM_011518285.1 | XP_011516587.1 | 12 | 1799 | |||
XM_011518288.1 | XP_011516590.1 | 11 | 1657 | |||
XM_011518289.2 | XP_011516591.1 | 14 | 1970 | |||
XM_011518290.1 | XP_011516592.1 | 13 | 1917 | |||
XM_011518291.1 | XP_011516593.1 | 13 | 1971 | |||
XM_011518292.1 | XP_011516594.1 | 12 | 1842 | |||
XM_011518293.1 | XP_011516595.1 | 14 | 2037 | |||
GenBank mRNA | AK057342.1 | - | 14 | 2071 | ||
AK074105.1 | - | 6 | 4403 | |||
AK128011.1 | - | 8 | 1641 | |||
AK302737.1 | - | 14 | 2032 | |||
BC031281.1 | - | 14 | 2084 | 5295890 |