Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145307.3 | NP_660350.2 | 10 | 1324 | ||
XM_011539456.2 | XP_011537758.1 | 12 | 1445 | |||
XM_011539457.2 | XP_011537759.1 | 12 | 1445 | |||
XM_011539460.2 | XP_011537762.1 | 12 | 1445 | |||
XM_017015842.1 | XP_016871331.1 | 11 | 1347 | |||
XM_017015843.1 | XP_016871332.1 | 12 | 1445 | |||
XM_017015844.1 | XP_016871333.1 | 10 | 1340 | |||
GenBank mRNA | AK096671.1 | - | 6 | 502 | ||
AY150309.1 | - | 10 | 1168 |