Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 23, 24 | 2749 | ||
NR_104452.1 | 26, 27 | 3147 | ||||
XM_005266861.2 | XP_005266918.1 | 24, 25 | 2872 | |||
XM_011535569.1 | XP_011533871.1 | 24, 25 | 2872 | |||
XM_011535570.1 | XP_011533872.1 | 25, 26 | 2920 | |||
XM_011535571.2 | XP_011533873.1 | 25, 26 | 3573 | |||
XM_011535574.1 | XP_011533876.1 | 24, 25 | 2868 | |||
XM_011535575.2 | XP_011533877.1 | 25, 26 | 2920 | |||
XM_011535576.2 | XP_011533878.1 | 25, 26 | 2920 | |||
XM_011535580.2 | XP_011533882.1 | 15, 16 | 1723 | |||
XM_017010397.1 | XP_016865886.1 | 25, 26 | 2920 | |||
XM_017010398.1 | XP_016865887.1 | 25, 26 | 3568 | |||
XM_017010399.1 | XP_016865888.1 | 24, 25 | 2797 | |||
XM_017010400.1 | XP_016865889.1 | 24, 25 | 2794 | |||
XM_017010401.1 | XP_016865890.1 | 24, 25 | 2797 | |||
XM_017010402.1 | XP_016865891.1 | 24, 25 | 2872 | |||
XM_017010403.1 | XP_016865892.1 | 25, 26 | 2920 | |||
XM_017010404.1 | XP_016865893.1 | 10, 11 | 1143 | |||
GenBank mRNA | AK055461.1 | - | 7, 8 | 694 | ||
AK091554.1 | - | 23, 24 | 2730 | |||
AK125385.1 | - | 13, 14 | 1582 | |||
AK126259.1 | - | 3, 4 | 318 | |||
AK131446.1 | - | 26, 27 | 3127 |