Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152730.5 | NP_689943.4 | 27 | 3088 | ||
NR_104452.1 | 31 | 4055 | ||||
XM_005266861.2 | XP_005266918.1 | 28 | 3211 | |||
XM_011535569.1 | XP_011533871.1 | 29 | 3274 | |||
XM_011535570.1 | XP_011533872.1 | 30 | 3322 | |||
XM_011535571.2 | XP_011533873.1 | 30 | 3975 | |||
XM_011535574.1 | XP_011533876.1 | 29 | 3270 | |||
XM_011535580.2 | XP_011533882.1 | 20 | 2125 | |||
XM_017010397.1 | XP_016865886.1 | 29 | 3259 | |||
XM_017010398.1 | XP_016865887.1 | 29 | 3907 | |||
XM_017010399.1 | XP_016865888.1 | 29 | 3199 | |||
XM_017010400.1 | XP_016865889.1 | 29 | 3196 | |||
XM_017010401.1 | XP_016865890.1 | 28 | 3136 | |||
XM_017010404.1 | XP_016865893.1 | 15 | 1545 | |||
GenBank mRNA | AK055461.1 | - | 11 | 1033 | ||
AK091554.1 | - | 27 | 3069 | |||
AK125385.1 | - | 17 | 1921 | |||
AK126259.1 | - | 7 | 657 | |||
AK131446.1 | - | 31 | 4035 |