Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153614.3 | NP_705842.2 | 8 | 1695 | ||
XM_005273984.3 | XP_005274041.1 | 7 | 1060 | |||
XM_011545004.2 | XP_011543306.1 | 10 | 1946 | |||
XM_011545005.2 | XP_011543307.1 | 9 | 1786 | |||
XM_011545007.2 | XP_011543309.1 | 8 | 1471 | |||
XM_011545009.2 | XP_011543311.1 | 10 | 1620 | |||
XM_011545015.2 | XP_011543317.1 | 5 | 813 | |||
XM_017017675.1 | XP_016873164.1 | 9 | 1216 | |||
XM_017017676.1 | XP_016873165.1 | 10 | 1796 | |||
XM_017017677.1 | XP_016873166.1 | 9 | 1665 | |||
XM_017017678.1 | XP_016873167.1 | 8 | 1476 | |||
GenBank mRNA | AF516185.1 | - | 5 | 1667 | ||
AK058095.1 | - | 5 | 1667 | |||
AK128589.1 | - | 10 | 3740 | |||
AY138810.2 | - | 8 | 1695 | |||
AY325766.1 | - | 8 | 988 |