Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL24D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016304.2 | NP_057388.1 | 6 | 575 | ||
GenBank mRNA | AB462502.1 | - | 6 | 459 | ||
AF060926.1 | - | 6 | 442 | |||
AF165521.1 | - | 6 | 575 | |||
AF201949.1 | - | 6 | 575 | |||
AF212226.1 | - | 6 | 575 | |||
AK055713.1 | - | 6 | 462 | |||
AK315431.1 | - | 6 | 475 | |||
BC008409.1 | - | 6 | 497 | 4097130 | ||
BC008422.1 | - | 6 | 467 | 4096889 | ||
BC008449.1 | - | 6 | 473 | 4133589 | ||
BC008499.1 | - | 6 | 488 | 4286115 | ||
BC009593.1 | - | 6 | 478 | 4109097 | ||
BC009604.1 | - | 6 | 478 | 4282059 | ||
BC012913.1 | - | 6 | 442 | 4514979 | ||
BC014576.1 | - | 6 | 477 | 3930303 | ||
BC016312.1 | - | 6 | 483 | 4096867 | ||
BC016331.1 | - | 6 | 483 | 4108672 | ||
BC016725.1 | - | 6 | 484 | 4078305 | ||
BC016777.1 | - | 6 | 465 | 4104810 | ||
BC026266.1 | - | 6 | 479 | 4276092 | ||
BC026267.1 | - | 6 | 458 | 4277062 | ||
BC035995.1 | - | 6 | 488 | 4671856 | ||
BC093067.1 | - | 6 | 462 | 30419021 | ||
CR457179.1 | - | 6 | 432 |