Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042415.2 | NP_001035880.1 | 6 | 611 | ||
NM_001042416.2 | NP_001035881.1 | 6 | 744 | |||
NM_001287254.1 | NP_001274183.1 | 6 | 602 | |||
NM_001287255.1 | NP_001274184.1 | 6 | 577 | |||
NM_001287256.1 | NP_001274185.1 | 6 | 573 | |||
NM_001287399.1 | NP_001274328.1 | 6 | 660 | |||
NM_173539.3 | NP_775810.2 | 6 | 662 | |||
XM_011534744.1 | XP_011533046.1 | 6 | 584 | |||
XM_017013166.1 | XP_016868655.1 | 5 | 2301 | |||
GenBank mRNA | AK313454.1 | - | 6 | 485 | ||
BC026190.1 | - | 6 | 485 | 4837032 | ||
BM478564.1 | - | 6 | 473 | |||
DB047692.1 | - | 6 | 494 | |||
DB055646.1 | - | 6 | 542 | |||
DN995476.1 | - | 6 | 606 |