Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM153C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038353.1 | 4, 5 | 238 | |||
GenBank mRNA | AK294762.1 | - | 4, 5 | 232 | ||
BC101338.1 | - | 3, 4 | 272 | 40025116 | ||
BC101339.2 | - | 5, 6 | 497 | |||
BC101340.2 | - | 3, 4 | 272 | |||
BC101341.2 | - | 3, 4 | 272 | 40025123 | ||
BC131737.1 | - | 3, 4 | 272 | 40025113 | ||
BC146883.1 | - | 3, 4 | 484 | 9056811 | ||
BC146888.1 | - | 3, 4 | 484 | 9056816 | ||
BC171810.1 | - | 3, 4 | 485 | 9054464 | ||
DB030341.1 | - | 4, 5 | 239 |
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265615.1 | NP_001252544.1 | 3, 4 | 143 | ||
GenBank mRNA | BC028606.1 | - | 4, 5 | 384 | 5266126 |