Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300834.1 | NP_001287763.1 | 2 | 201 | ||
NM_001300835.1 | NP_001287764.1 | 2 | 201 | |||
NM_174896.3 | NP_777556.1 | 2 | 201 | |||
XM_011540712.2 | XP_011539014.1 | 3 | 5681 | |||
XM_017000323.1 | XP_016855812.1 | 3 | 1550 | |||
XM_017000324.1 | XP_016855813.1 | 3 | 5681 | |||
GenBank mRNA | AK123160.1 | - | 1 | 1489 | ||
BC017973.1 | - | 2 | 88 | 4693393 | ||
BG545926.1 | - | 2 | 91 | |||
HX785231.1 | - | 2 | 83 |