Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBSCR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182504.3 | NP_872310.2 | 2 | 421 | ||
XM_011515785.2 | XP_011514087.1 | 2 | 355 | |||
XM_017011741.1 | XP_016867230.1 | 2 | 543 | |||
GenBank mRNA | AY372053.1 | - | 2 | 426 | ||
AY372054.1 | - | 3 | 472 | |||
BC030643.2 | - | 2 | 421 | 4824662 |