Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182526.2 | NP_872332.1 | 3 | 654 | ||
XM_005267373.2 | XP_005267430.1 | 3 | 660 | |||
XM_005267374.3 | XP_005267431.1 | 4 | 527 | |||
XM_005267375.2 | XP_005267432.1 | 3 | 719 | |||
XM_006720062.2 | XP_006720125.1 | 3 | 539 | |||
XM_017021051.1 | XP_016876540.1 | 4 | 811 | |||
XM_017021052.1 | XP_016876541.1 | 3 | 882 | |||
XM_017021053.1 | XP_016876542.1 | 3 | 786 | |||
XM_017021054.1 | XP_016876543.1 | 3 | 1024 | |||
XM_017021055.1 | XP_016876544.1 | 3 | 946 | |||
GenBank mRNA | AK090706.1 | - | 3 | 476 | ||
BC104940.1 | - | 2 | 390 | 8143943 | ||
BC104942.1 | - | 2 | 390 | 8143945 |