Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC25A45 SIRNA Assays ›
Gene Symbol:
SLC25A45
Gene Name:
solute carrier family 25 member 45
Gene Aliases:
-
Chromosome Location:
Chr.11: 65375192 - 65383701 on Build GRCh38
UniGene ID:
Hs.661604
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
242130
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC25A45
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077241.2
NP_001070709.2
2
221
NM_001278250.2
NP_001265179.2
2
504
NM_001278251.2
NP_001265180.2
6
1078
NM_001300820.1
NP_001287749.1
2
504
NM_182556.3
NP_872362.3
3
276
XM_006718509.3
XP_006718572.1
2
886
XM_006718510.3
XP_006718573.1
4
414
XM_011544943.2
XP_011543245.1
3
985
XM_011544944.2
XP_011543246.1
2
315
XM_011544947.2
XP_011543249.1
2
504
XM_011544949.2
XP_011543251.1
4
333
XM_017017562.1
XP_016873051.1
2
504
XM_017017563.1
XP_016873052.1
3
1751
XM_017017564.1
XP_016873053.1
3
1750
XM_017017565.1
XP_016873054.1
3
1015
XM_017017566.1
XP_016873055.1
3
370
XM_017017567.1
XP_016873056.1
4
376
XM_017017568.1
XP_016873057.1
3
199
XM_017017569.1
XP_016873058.1
5
1078
XM_017017570.1
XP_016873059.1
2
887
XM_017017571.1
XP_016873060.1
2
257
GenBank mRNA
AK091190.1
-
6
1078
AY597807.1
-
2
69
BC036869.1
-
2
184
5228549
BX648573.1
-
1
2503
DA453096.1
-
2
504
DA872876.1
-
3
273
DB049668.1
-
3
277
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
translation
transmembrane transport
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0