Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182578.3 | NP_872384.1 | 1, 2 | 253 | ||
XM_011509421.1 | XP_011507723.1 | 1, 2 | 253 | |||
GenBank mRNA | AK095283.1 | - | 1, 2 | 253 | ||
BC101610.1 | - | 1, 2 | 159 | 8069116 | ||
BC112239.1 | - | 1, 2 | 159 | 8327707 |