Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016333.3 | NP_057417.3 | 3 | 806 | ||
XM_005255226.1 | XP_005255283.1 | 3 | 784 | |||
XM_006720874.3 | XP_006720937.1 | 3 | 784 | |||
GenBank mRNA | AB002322.3 | - | 3 | 458 | ||
AB016088.1 | - | 2 | 154 | |||
AB016089.1 | - | 3 | 442 | |||
AB016092.1 | - | 3 | 482 | |||
AF201422.1 | - | 3 | 486 | |||
BC007752.2 | - | 3 | 458 | |||
BC020639.1 | - | 3 | 497 | |||
BC041155.1 | - | 2 | 187 | |||
BC046104.1 | - | 3 | 462 | |||
BC070050.1 | - | 3 | 517 | |||
BX647088.1 | - | 2 | 352 |