Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100287896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319240.1 | NP_001306169.1 | 2 | 1269 | ||
XM_017017055.1 | XP_016872544.1 | 2 | 757 | |||
GenBank mRNA | BC019340.1 | - | 2 | 1214 | 3051084 | |
BX647206.1 | - | 2 | 858 |