Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 55 | 8339 | ||
XM_006712699.3 | XP_006712762.1 | 56 | 8452 | |||
GenBank mRNA | AB053319.1 | - | 5 | 590 | ||
AK127772.1 | - | 28 | 3937 | |||
AK128636.1 | - | 25 | 3066 | |||
AY172970.1 | - | 26 | 3245 | |||
BX647133.1 | - | 19 | 2166 |