Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY230136.1 | - | 1 | 465 | ||
BC062480.1 | - | 1 | 658 | 30398001 | ||
BC125145.1 | - | 1 | 477 | 40120129 |
Gene Symbol: DPPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199286.3 | NP_954980.1 | 4 | 526 | ||
GenBank mRNA | AY317075.1 | - | 4 | 526 |