Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199341.3 | NP_955373.3 | 4 | 1754 | ||
XM_005259449.3 | XP_005259506.1 | 4 | 2192 | |||
XM_006723204.3 | XP_006723267.1 | 4 | 1889 | |||
XM_006723205.2 | XP_006723268.1 | 4 | 1780 | |||
XM_011526936.2 | XP_011525238.1 | 3 | 1794 | |||
XM_011526937.2 | XP_011525239.1 | 2 | 1296 | |||
GenBank mRNA | BC043386.1 | - | 4 | 1754 | 5492230 |