Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256477.1 | NP_001243406.1 | 1, 2 | 806 | ||
NM_001256478.1 | NP_001243407.1 | 1, 2 | 806 | |||
NM_207315.3 | NP_997198.2 | 1, 2 | 806 | |||
GenBank mRNA | AK127983.1 | - | 1, 2 | 894 | ||
BC089425.1 | - | 1, 2 | 659 | |||
BC132821.1 | - | 1, 2 | 806 | 40146843 | ||
BC141802.1 | - | 1, 2 | 699 | 40147165 | ||
BC144202.1 | - | 1, 2 | 806 | 9052722 |