Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321525.1 | NP_001308454.1 | 22 | 2899 | ||
NM_001321526.1 | NP_001308455.1 | 22 | 2989 | |||
NM_001321527.1 | NP_001308456.1 | 22 | 2953 | |||
NM_001321528.1 | NP_001308457.1 | 22 | 2943 | |||
NM_001321529.1 | NP_001308458.1 | 21 | 2776 | |||
NM_001321530.1 | NP_001308459.1 | 21 | 2668 | |||
NM_001321531.1 | NP_001308460.1 | 21 | 2689 | |||
NM_207328.3 | NP_997211.2 | 22 | 2971 | |||
XM_005263888.3 | XP_005263945.1 | 20 | 2393 | |||
XM_006712311.3 | XP_006712374.1 | 21 | 2543 | |||
XM_006712313.3 | XP_006712376.1 | 20 | 2411 | |||
XM_017003424.1 | XP_016858913.1 | 22 | 2603 | |||
XM_017003426.1 | XP_016858915.1 | 19 | 2333 | |||
GenBank mRNA | AK122580.1 | - | 12 | 4395 | ||
AK122734.1 | - | 14 | 1711 | |||
AK131194.1 | - | 21 | 2689 | |||
AK131197.1 | - | 16 | 2008 | |||
AK298131.1 | - | 21 | 2348 | |||
AK302887.1 | - | 20 | 3703 | |||
BC068596.1 | - | 22 | 2642 | 4837765 |