Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 22, 23 | 4135 | ||
NM_016195.3 | NP_057279.2 | 22, 23 | 4015 | |||
GenBank mRNA | AB033337.1 | - | 22, 23 | 4088 | ||
AK024959.1 | - | 5, 6 | 1710 | |||
AK025628.1 | - | 2, 3 | 170 | |||
AL117496.1 | - | 22, 23 | 3970 | |||
AL137392.1 | - | 9, 10 | 2602 | |||
AY282407.1 | - | 7, 8 | 1932 | |||
AY739715.1 | - | 3, 4 | 728 | |||
L16782.1 | - | 3, 4 | 282 | |||
U93121.1 | - | 3, 4 | 843 |