Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113567.2 | NP_001107039.1 | 4 | 2715 | ||
NM_207387.3 | NP_997270.2 | 3 | 2599 | |||
XM_011523845.2 | XP_011522147.1 | 5 | 2788 | |||
XM_017024619.1 | XP_016880108.1 | 4 | 2484 | |||
XM_017024620.1 | XP_016880109.1 | 5 | 2647 | |||
GenBank mRNA | AK093015.1 | - | 3 | 2258 | ||
AL833245.1 | - | 3 | 2625 | |||
AL833260.1 | - | 3 | 2625 |