Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMOTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278683.1 | NP_001265612.1 | 10 | 3188 | ||
NM_001278685.1 | NP_001265614.1 | 10 | 3196 | |||
NM_016201.3 | NP_057285.3 | 10 | 3205 | |||
XM_006713654.2 | XP_006713717.1 | 10 | 3220 | |||
XM_011512881.1 | XP_011511183.1 | 10 | 3059 | |||
XM_017006580.1 | XP_016862069.1 | 10 | 3062 | |||
XM_017006581.1 | XP_016862070.1 | 10 | 3243 | |||
XM_017006582.1 | XP_016862071.1 | 10 | 3070 | |||
GenBank mRNA | AB023206.1 | - | 10 | 3138 | ||
AF175966.1 | - | 9 | 2269 | |||
AK074731.1 | - | 5 | 1603 | |||
AK091040.1 | - | 7 | 2273 | |||
BC011454.1 | - | 8 | 2168 | 4181482 | ||
BC025981.2 | - | 7 | 1758 |