Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032296.3 | NP_001027467.2 | 11 | 2404 | ||
NM_001286649.1 | NP_001273578.1 | 10 | 2347 | |||
NM_003576.4 | NP_003567.2 | 11 | 2348 | |||
XM_005254079.4 | XP_005254136.1 | 11 | 2225 | |||
XM_017020794.1 | XP_016876283.1 | 11 | 2521 | |||
XM_017020795.1 | XP_016876284.1 | 11 | 2183 | |||
GenBank mRNA | AF083420.1 | - | 11 | 2221 | ||
BC065378.1 | - | 11 | 2348 | 6598075 | ||
EU668350.1 | - | 11 | 2395 |