Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 2 | 742 | ||
NR_036641.1 | 3 | 803 | ||||
XM_011532124.2 | XP_011530426.1 | 4 | 422 | |||
XM_017008455.1 | XP_016863944.1 | 2 | 371 | |||
XM_017008456.1 | XP_016863945.1 | 2 | 704 | |||
GenBank mRNA | AB033831.1 | - | 2 | 577 | ||
AF091434.1 | - | 2 | 742 | |||
AF244813.1 | - | 2 | 439 | |||
AF260738.1 | - | 2 | 413 | |||
AK313817.1 | - | 2 | 472 | |||
AM922296.1 | - | 3 | 623 | |||
AY358493.1 | - | 2 | 535 | |||
BC094746.1 | - | 2 | 343 | |||
BC136662.1 | - | 2 | 663 | 9020652 |