Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014796.1 | NP_001014796.1 | 9 | 1301 | ||
NM_006182.2 | NP_006173.2 | 8 | 1221 | |||
XM_006711344.3 | XP_006711407.1 | 8 | 1262 | |||
XM_011509586.2 | XP_011507888.1 | 8 | 1194 | |||
XM_011509587.2 | XP_011507889.1 | 9 | 1255 | |||
XM_011509588.2 | XP_011507890.1 | 8 | 1258 | |||
GenBank mRNA | AK095975.1 | - | 9 | 1274 | ||
AK225063.1 | - | 8 | 1140 | |||
AK225196.1 | - | 8 | 1196 | |||
AK314388.1 | - | 8 | 1170 | |||
BC052998.2 | - | 9 | 1272 | 6162716 | ||
BX640899.1 | - | 9 | 1258 | |||
X74764.1 | - | 8 | 1189 |