Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000954.5 | NP_000945.3 | 5, 6 | 635 | ||
GenBank mRNA | AK075333.1 | - | 5, 6 | 635 | ||
AK312817.1 | - | 5, 6 | 622 | |||
AY026356.1 | - | 5, 6 | 560 | |||
BC005939.1 | - | 5, 6 | 569 | 4294999 | ||
BC041463.1 | - | 7, 8 | 1843 | |||
BM686612.1 | - | 5, 6 | 623 | |||
BT019921.1 | - | 5, 6 | 560 | |||
BT019922.1 | - | 5, 6 | 560 | |||
M61900.2 | - | 5, 6 | 625 |